missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse Kirrel2/NEPH3 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£170.00 - £425.00
Specifications
| Antigen | Kirrel2/NEPH3 |
|---|---|
| Clone | 318311 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18772779
![]() |
R&D Systems
MAB2564-SP |
25 μg |
£170.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18131361
|
R&D Systems
MAB2564 |
100 μg |
£425.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Kirrel2/NEPH3 Monoclonal specifically detects Kirrel2/NEPH3 in Human, Mouse samples. It is validated for Western Blot.Specifications
| Kirrel2/NEPH3 | |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Mouse | |
| Q6UWL6 | |
| 84063 | |
| Mouse myeloma cell line NS0-derived recombinant human Kirrel2 Gly21-Arg503 Accession # Q6UWL6 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human and mouse Kirrel2/NEPH3 in direct ELISAs and Western blots. |
| 318311 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| DKFZp564A1164, Filtrin, FILTRINkin of IRRE-like protein 2, kin of IRRE like 2 (Drosophila), Kin of irregular chiasm-like protein 2, MGC15718, NEPH3, NEPH3Nephrin-like protein 3, nephrin-like 3, nephrin-like gene 1, NLG1, NLG1kin of irregular chiasm-like 2 | |
| KIRREL2 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title