missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse Kirrel2/NEPH3 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB2564
This item is not returnable.
View return policy
Description
Kirrel2/NEPH3 Monoclonal specifically detects Kirrel2/NEPH3 in Human, Mouse samples. It is validated for Western Blot.
Specifications
| Kirrel2/NEPH3 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q6UWL6 | |
| KIRREL2 | |
| Mouse myeloma cell line NS0-derived recombinant human Kirrel2 Gly21-Arg503 Accession # Q6UWL6 | |
| 100 μg | |
| Primary | |
| Detects human and mouse Kirrel2/NEPH3 in direct ELISAs and Western blots. | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| 318311 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| DKFZp564A1164, Filtrin, FILTRINkin of IRRE-like protein 2, kin of IRRE like 2 (Drosophila), Kin of irregular chiasm-like protein 2, MGC15718, NEPH3, NEPH3Nephrin-like protein 3, nephrin-like 3, nephrin-like gene 1, NLG1, NLG1kin of irregular chiasm-like 2 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 84063 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction