missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kir6.1 Antibody (S366-60), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
£181.00 - £456.00
Specifications
| Antigen | Kir6.1 |
|---|---|
| Clone | S366-60 |
| Dilution | Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18290155
|
Novus Biologicals
NBP2-59324 |
100 μg |
£456.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18668857
|
Novus Biologicals
NBP2-59324-25ug |
25 μg |
£181.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Kir6.1 Monoclonal specifically detects Kir6.1 in Human, Mouse, Rat, Gerbil samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
| Kir6.1 | |
| Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat, Gerbil | |
| 3764 | |
| Fusion protein amino acids 306-424 (Cytoplasmic C-terminus) of rat Kir6.1 | |
| Primary | |
| Detects 45kDa. |
| S366-60 | |
| Monoclonal | |
| Purified | |
| RUO | |
| ATP-sensitive inward rectifier potassium channel 8, Inward rectifier K(+) channel Kir6.1, inwardly rectifying potassium channel KIR6.1, KIR6.1, Potassium channel, inwardly rectifying subfamily J member 8, potassium inwardly-rectifying channel, subfamily J, member 8, uKATP-1 | |
| KCNJ8 | |
| IgG2a | |
| Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title