missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kir6.1 Antibody (S366-60), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP2-59324
This item is not returnable.
View return policy
Description
Kir6.1 Monoclonal specifically detects Kir6.1 in Human, Mouse, Rat, Gerbil samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| Kir6.1 | |
| Monoclonal | |
| Unconjugated | |
| PBS pH7.4, 50% glycerol with 0.1% Sodium Azide | |
| KCNJ8 | |
| Fusion protein amino acids 306-424 (Cytoplasmic C-terminus) of rat Kir6.1 | |
| 100 μg | |
| Primary | |
| Detects 45kDa. | |
| Store at -20C. | |
| IgG2a |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| S366-60 | |
| Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 | |
| ATP-sensitive inward rectifier potassium channel 8, Inward rectifier K(+) channel Kir6.1, inwardly rectifying potassium channel KIR6.1, KIR6.1, Potassium channel, inwardly rectifying subfamily J member 8, potassium inwardly-rectifying channel, subfamily J, member 8, uKATP-1 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 3764 | |
| Human, Mouse, Rat, Gerbil | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction