missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIR2DS4/CD158i Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£359.00
Specifications
| Antigen | KIR2DS4/CD158i |
|---|---|
| Dilution | Western Blot 1.0 ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
KIR2DS4/CD158i Polyclonal specifically detects KIR2DS4/CD158i in Human samples. It is validated for Western Blot.Specifications
| KIR2DS4/CD158i | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| CD158 antigen-like family member I, CD158I, CD158i antigen, cl-39, killer cell immunoglobulin-like receptor 2DS4, killer cell immunoglobulin-like receptor, two domains, short cytoplasmic tail4,nkat8, killer inhibitory receptor 4-1-2, KIR antigen 2DS4, KKA3KIR1D, MGC120019, MGC125315, MGC125317, MHC class I NK cell receptor, natural killer cell inhibitory receptor, Natural killer-associated transcript 8, NKAT-8, NKAT8KIR412, P58 natural killer cell receptor clones CL-39/CL-17, p58 NK receptor CL-39/CL-17 | |
| The immunogen for Anti-KIR2DS4/CD158i antibody is: synthetic peptide directed towards the C-terminal region of Human KI2S4 (NP_036446.3). Peptide sequence RDAPYEWSNSSDPLLVSVTGNPSNSWPSPTEPSSKTGNPRHLHVLIGTSV | |
| Affinity purified |
| Western Blot 1.0 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS buffer, 2% sucrose | |
| 3809 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title