missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kir2.1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | Kir2.1 |
|---|---|
| Concentration | 0.33 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16879444
|
Proteintech
19965-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16869444
|
Proteintech
19965-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The antibody recognizes the C-term of KCNJ2.
The KIR2 subunit family includes 2.1, 2.2, 2.3 and 2.4. Unlike G protein-coupled KIR3 subunits, KIR2.1 requires both phosphorylation by PKA and ATP hydrolysis for functional activity. KIR2.1 is expressed in the superior and inferior collicula and the pontine region of the brain, where it moderates synaptic transmission, like many other potassium channels. In the placenta, KIR2.1 is expressed throughout gestation in cytotrophoblast cells. In the kidney, KIR2.1 colocalizes with KIR5.1 in the proximal tubule. KIR2.1, 2.2 and 2.3 associate with the membrane-associated guanylate kinase synapseassociated protein 97 in the cerebellum and heart. Phosphorylation of KIR2.2 by protein kinase A inhibits the associates with SAP97.Specifications
| Kir2.1 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| Mouse, Rat, Human | |
| P35561, P63252, Q64273 | |
| 16518, 29712, 3759 | |
| Peptide | |
| Primary | |
| -20°C | |
| KCNJ2 |
| 0.33 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| HHBIRK1, HHIRK1, hIRK1, IRK 1, IRK1, KCNJ2, Kir2.1, LQT7, SQT3 | |
| Kcnj2 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title