missing translation for 'onlineSavingsMsg'
Learn More

Kir2.1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 16879444 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16879444 20 μL 20µL
16869444 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16879444 Supplier Proteintech Supplier No. 199651AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The antibody recognizes the C-term of KCNJ2.

The KIR2 subunit family includes 2.1, 2.2, 2.3 and 2.4. Unlike G protein-coupled KIR3 subunits, KIR2.1 requires both phosphorylation by PKA and ATP hydrolysis for functional activity. KIR2.1 is expressed in the superior and inferior collicula and the pontine region of the brain, where it moderates synaptic transmission, like many other potassium channels. In the placenta, KIR2.1 is expressed throughout gestation in cytotrophoblast cells. In the kidney, KIR2.1 colocalizes with KIR5.1 in the proximal tubule. KIR2.1, 2.2 and 2.3 associate with the membrane-associated guanylate kinase synapseassociated protein 97 in the cerebellum and heart. Phosphorylation of KIR2.2 by protein kinase A inhibits the associates with SAP97.

Specifications

Antigen Kir2.1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.33 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene KCNJ2
Gene Accession No. P35561, P63252, Q64273
Gene Alias HHBIRK1, HHIRK1, hIRK1, IRK 1, IRK1, KCNJ2, Kir2.1, LQT7, SQT3
Gene Symbols Kcnj2
Host Species Rabbit
Immunogen Peptide
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16518, 29712, 3759
Target Species Mouse, Rat, Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.