missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Kinase Tracer 314, 25 μM

Product Code. 10458468
Change view
Click to view available options
Quantity:
25 μL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10458468 25 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10458468 Supplier Invitrogen™ Supplier No. PV6087
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Alexa Fluor™ Kinase Tracer 315 is detected by addition of a Eu-labeled anti-tag antibody.

  • Excitation/Emission (nm): 340/615, 665
  • Label or Dye: Alexa Fluor™ 647
  • Target Enzyme: Kinases (Multiple)
  • Validated Kinases: PIK3CD, PIK3CG, FRAP1 (mTOR)
  • In the LanthaScreen™ Eu Kinase Binding Assay, binding of an Alexa Fluor™ conjugate or tracer such as Kinase Tracer 314 to a kinase is detected by addition of a Eu-labeled anti-tag antibody. Binding of the tracer & antibody to a kinase results in a high degree of FRET, whereas displacement of the tracer with a kinase inhibitor results in a loss of FRET. Unlike many kinase activity assays, this assay is a simple mix and read assay, with no development steps.
TRUSTED_SUSTAINABILITY

Specifications

Assay Kinase Binding Assay
Assay Entry Biochemical binding/kinetic
Content And Storage Store at -20°C.
Excitation/Emission 340/615, 665
For Use With (Application) TR-FRET
For Use With (Equipment) Microplate Reader
Format 384-well Plate
Label or Dye Alexa Fluor™ 647
Quantity 25 μL
Readout Kinetic
Shipping Condition Dry Ice
Target Entry PIK3CG, PIK3CD/PIK3R1, mTOR, FRAP1
Type Kinase Tracer
Validated Kinases PIK3CD, PIK3CG, FRAP1 (mTOR)
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.