missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Kimble™ KIMAX™ Colorware™ Low-Form Beakers, red

Kimax-33 glass beakers with brightly colored graduations and marking spot

Specifications

Color Raging Red
Material Borosilicate Glass
For Use With (Application) For Easy Identification in The Lab, Reduce Cross Contamination
Label Type Matte Finish Marking
Type Beaker
View More Specs

Description

Description

  • With pouring spout
  • Matte-finish marking spot can be marked with ordinary pencil
  • Colors make graduated scale easy to read

Compliance

Designed from ASTM™ E 960 Type I requirements

Specifications

Specifications

Raging Red
For Easy Identification in The Lab, Reduce Cross Contamination
Beaker
ASTM E960, Type I
Borosilicate Glass
Matte Finish Marking
Yes
Dual Scale
Videos
SDS
Documents

Documents

Product Certifications