missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIF21A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£222.00 - £513.00
Specifications
| Antigen | KIF21A |
|---|---|
| Dilution | Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:20 - 1:50 |
| Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18613336
|
Novus Biologicals
NBP2-37969-25ul |
25 μL |
£222.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18199377
|
Novus Biologicals
NBP2-37969 |
0.1 mL |
£513.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
KIF21A Polyclonal specifically detects KIF21A in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| KIF21A | |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| CFEOM1, CFEOM3B, FEOM1, FEOM3A, fibrosis of the extraocular muscles, congenital, 1, FLJ20052, KIAA1708DKFZp779C159, KIF2, kinesin family member 21A, Kinesin-like protein KIF2, kinesin-like protein KIF21A, Renal carcinoma antigen NY-REN-62 | |
| KIF21A | |
| IgG | |
| Affinity Purified |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| Q7Z4S6 | |
| 55605 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: QKEAQIKVLEGRLKQTEITSATQNQLLFHMLKEKAELNPELDALLGHALQDLDSVPLENVEDSTDEDAPLNS | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title