Learn More
KIF20A Mouse anti-Human, Unconjugated, Clone: 2A8E3, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67190-1-IG-150UL
Description
The KIF20A antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human KIF20A. This antibody recognizes human, rat, mouse antigen. The KIF20A antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
| KIF20A | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Rat, Mouse |
| Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
| 2A8E3 | |
| Unconjugated | |
| 150 μL | |
| 10112 | |
| IgG1 |