missing translation for 'onlineSavingsMsg'
Learn More

Khasianine, TRC

Product Code. 30391280
Change view
Click to view available options
Quantity:
2.5 mg
5 mg
25 mg
Unit Size:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30391280 5 mg -
30396543 25 mg -
30396190 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30391280 Supplier Toronto Research Chemicals Supplier No. TRCK5200005MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32449-98-2
Molecular Formula C24 H26 O5 S
Molecular Weight (g/mol) 426.525
Synonym (3β,22α,25R)-Spirosol-5-en-3-yl 4-O-(6-deoxy-α-L-mannopyranosyl)-β-D-glucopyranoside, Khasianine, NSC 376459, β2-Solamargine
IUPAC Name [4-[2-[4-[2-(4-hydroxyphenyl)propan-2-yl]phenyl]propan-2-yl]phenyl] hydrogen sulfate
SMILES CC(C)(c1ccc(O)cc1)c2ccc(cc2)C(C)(C)c3ccc(OS(=O)(=O)O)cc3

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C24H26O5S/c1-23(2,19-9-13-21(25)14-10-19)17-5-7-18(8-6-17)24(3,4)20-11-15-22(16-12-20)29-30(26,27)28/h5-16,25H,1-4H3,(H,26,27,28)
Formula Weight 426.15
Chemical Name or Material Khasianine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.