CAS RN 32449-98-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Khasianine, TRC
CAS: 32449-98-2 Molecular Formula: C24 H26 O5 S Molecular Weight (g/mol): 426.525 Synonym: (3β,22α,25R)-Spirosol-5-en-3-yl 4-O-(6-deoxy-α-L-mannopyranosyl)-β-D-glucopyranoside,Khasianine,NSC 376459,β2-Solamargine IUPAC Name: [4-[2-[4-[2-(4-hydroxyphenyl)propan-2-yl]phenyl]propan-2-yl]phenyl] hydrogen sulfate SMILES: CC(C)(c1ccc(O)cc1)c2ccc(cc2)C(C)(C)c3ccc(OS(=O)(=O)O)cc3