missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KGA/GAC Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£130.00 - £339.00
Specifications
Antigen | KGA/GAC |
---|---|
Concentration | 0.23 mg/mL |
Applications | Western Blot, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16870794
|
Proteintech
12855-1-AP-20UL |
20 μL |
£130.00
20µL |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16860794
|
Proteintech
12855-1-AP-150UL |
150 μL |
£339.00
150µL |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This antibody can recognize KGA and GAC.
Sahai (1983) demonstrated phosphate-activated glutaminase (EC 3.5.1.2) in human platelets. It is the major enzyme yielding glutamate from glutamine. Significance of the enzyme derives from its possible implication in behavior disturbances in which glutamate acts as a neurotransmitter(Prusiner, 1981). High heritability of platelet glutaminase was indicated by studies of Sahai and Vogel (1983) who found an intraclass correlation coefficient of 0.96 for monozygotic twins and 0.53 for dizygotic twins.Specifications
KGA/GAC | |
Western Blot, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunofluorescence | |
Unconjugated | |
Rabbit | |
Rat, Human, Mouse | |
D3Z7P3, O94925, P13264 | |
14660, 24398, 2744 | |
KGA/GAC Fusion Protein Ag3505 | |
Primary | |
-20°C | |
GLS |
0.23 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
AAD20, GAC, GLS, GLS1, glutaminase, K glutaminase, KGA, KIAA0838, L glutamine amidohydrolase | |
GLS | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |