missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KGA/GAC Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12855-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
This antibody can recognize KGA and GAC.
Sahai (1983) demonstrated phosphate-activated glutaminase (EC 3.5.1.2) in human platelets. It is the major enzyme yielding glutamate from glutamine. Significance of the enzyme derives from its possible implication in behavior disturbances in which glutamate acts as a neurotransmitter(Prusiner, 1981). High heritability of platelet glutaminase was indicated by studies of Sahai and Vogel (1983) who found an intraclass correlation coefficient of 0.96 for monozygotic twins and 0.53 for dizygotic twins.Specifications
KGA/GAC | |
Polyclonal | |
Unconjugated | |
GLS | |
AAD20, GAC, GLS, GLS1, glutaminase, K glutaminase, KGA, KIAA0838, L glutamine amidohydrolase | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
14660, 24398, 2744 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunofluorescence | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
D3Z7P3, O94925, P13264 | |
GLS | |
KGA/GAC Fusion Protein Ag3505 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |