missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KF1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-31700
This item is not returnable.
View return policy
Description
KF1 Polyclonal specifically detects KF1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| KF1 | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| O00237 | |
| RNF103 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: YFEKKRRRNNNNDEVNANNLEWLSSLWDWYTSYLFHPIASFQNFPVESDWDEDPDLFLERLAFPDLWLHPLIPTDYIKNLPMWRFKCLGVQSE | |
| 0.1 mL | |
| Zinc Finger | |
| 7844 | |
| Human | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| E3 ubiquitin-protein ligase RNF103, EC 6.3.2.-, hkf-1, KF-1, KF1ZFP103ZFP-103, MGC102815, ring finger protein 103MGC41857, Zfp-103, Zinc finger protein 103 homolog, zinc finger protein 103 homolog (mouse), zinc finger protein expressed in cerebellum | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction