Learn More
Ketotifen fumarate, 99%, Thermo Scientific™
£41.90 - £132.00
Chemical Identifiers
| CAS | 34580-14-8 |
|---|---|
| Molecular Formula | C23H23NO5S |
| Molecular Weight (g/mol) | 425.499 |
| MDL Number | MFCD00079394 |
| InChI Key | YNQQEYBLVYAWNX-WLHGVMLRSA-N |
| Synonym | ketotifen fumarate, zaditen, ketotifen hydrogen fumarate, alaway, unii-hbd503woro, hc 20,511 fumarate, ketotifen fumarate usan:jan, ketotifen fumarate salt, hbd503woro |
| PubChem CID | 5282408 |
| IUPAC Name | (E)-but-2-enedioic acid;10-(1-methylpiperidin-4-ylidene)-5H-benzo[1,2]cyclohepta[3,4-b]thiophen-4-one |
| SMILES | CN1CCC(=C2C3=C(C(=O)CC4=CC=CC=C42)SC=C3)CC1.C(=CC(=O)O)C(=O)O |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15485889
|
Thermo Scientific Alfa Aesar
J63708.MD |
250mg |
£41.90
250mg |
Estimated Shipment: 12-10-2026
Log in to see stock. |
|||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
|
15475889
|
Thermo Scientific Alfa Aesar
J63708.03 |
1g |
£132.00
1g |
Estimated Shipment: 05-10-2026
Log in to see stock. |
|||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
SolubilitySoluble in water (10 mg/ml), DMSO (100 mM), and methanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Chemical Identifiers
| 34580-14-8 | |
| 425.499 | |
| YNQQEYBLVYAWNX-WLHGVMLRSA-N | |
| 5282408 | |
| CN1CCC(=C2C3=C(C(=O)CC4=CC=CC=C42)SC=C3)CC1.C(=CC(=O)O)C(=O)O |
| C23H23NO5S | |
| MFCD00079394 | |
| ketotifen fumarate, zaditen, ketotifen hydrogen fumarate, alaway, unii-hbd503woro, hc 20,511 fumarate, ketotifen fumarate usan:jan, ketotifen fumarate salt, hbd503woro | |
| (E)-but-2-enedioic acid;10-(1-methylpiperidin-4-ylidene)-5H-benzo[1,2]cyclohepta[3,4-b]thiophen-4-one |
Specifications
| 34580-14-8 | |
| MFCD00079394 | |
| ketotifen fumarate, zaditen, ketotifen hydrogen fumarate, alaway, unii-hbd503woro, hc 20,511 fumarate, ketotifen fumarate usan:jan, ketotifen fumarate salt, hbd503woro | |
| YNQQEYBLVYAWNX-WLHGVMLRSA-N | |
| (E)-but-2-enedioic acid;10-(1-methylpiperidin-4-ylidene)-5H-benzo[1,2]cyclohepta[3,4-b]thiophen-4-one | |
| 5282408 | |
| 99% |
| C23H23NO5S | |
| 14,5307 | |
| Soluble in water (10mg/ml),DMSO (100 mM),and methanol. | |
| CN1CCC(=C2C3=C(C(=O)CC4=CC=CC=C42)SC=C3)CC1.C(=CC(=O)O)C(=O)O | |
| 425.499 | |
| 425.5 | |
| Ketotifen fumarate |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
missing translation for 'einecsNumber' : 252-100-0
missing translation for 'rtecsNumber' : DE8260000
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.