missing translation for 'onlineSavingsMsg'
Learn More

Ketotifen fumarate, 99%, Thermo Scientific™

Brand:  Thermo Scientific Alfa Aesar J63708.03

Quantity 1g

 View more versions of this product

Product Code. 15475889

  • £132.00 / 1g
 Restricted Item
Estimated Shipment: 05-10-2026
This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Solubility
Soluble in water (10 mg/ml), DMSO (100 mM), and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

34580-14-8
425.499
YNQQEYBLVYAWNX-WLHGVMLRSA-N
5282408
CN1CCC(=C2C3=C(C(=O)CC4=CC=CC=C42)SC=C3)CC1.C(=CC(=O)O)C(=O)O
C23H23NO5S
MFCD00079394
ketotifen fumarate, zaditen, ketotifen hydrogen fumarate, alaway, unii-hbd503woro, hc 20,511 fumarate, ketotifen fumarate usan:jan, ketotifen fumarate salt, hbd503woro
(E)-but-2-enedioic acid;10-(1-methylpiperidin-4-ylidene)-5H-benzo[1,2]cyclohepta[3,4-b]thiophen-4-one

Specifications

34580-14-8
MFCD00079394
14,5307
Soluble in water (10mg/ml),DMSO (100 mM),and methanol.
CN1CCC(=C2C3=C(C(=O)CC4=CC=CC=C42)SC=C3)CC1.C(=CC(=O)O)C(=O)O
425.499
425.5
Ketotifen fumarate
C23H23NO5S
1g
ketotifen fumarate, zaditen, ketotifen hydrogen fumarate, alaway, unii-hbd503woro, hc 20,511 fumarate, ketotifen fumarate usan:jan, ketotifen fumarate salt, hbd503woro
YNQQEYBLVYAWNX-WLHGVMLRSA-N
(E)-but-2-enedioic acid;10-(1-methylpiperidin-4-ylidene)-5H-benzo[1,2]cyclohepta[3,4-b]thiophen-4-one
5282408
99%
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.