Learn More
DWK Life Sciences KECK™ Tubing Clamp, PBT, KT
For infinitely variable regulation of flow rates.
Brand: DWK Life Sciences 286338401
Code : A0
Additional Details : Weight : 0.01800kg
Description
With just one finger, flow rates in hoses can be infinitely adjusted with KECK™ hose clamps. The clamps are available in different colours for the different hose diameters.
- Continuous regulation of flow rates
- Handling with one hand possible
- Color coding of the clamps according to the different hose diameters
Specifications
100 | |
Infinitely Variable Regulation of Flow Rates | |
Fits tube OD 14 mm |
PBT | |
infinitely variable regulation of flow rates | |
Red |