Learn More
DWK Life Sciences KECK™ Clip, for conical joints, from POM
The KECK™ clamp is used to secure a standard ground joint connection.
Brand:  DWK Life Sciences 1091144
Code : NEW
Additional Details : Weight : 0.02000kg
 View more versions of this product
Product Code. 10264632
  • £32.80 / Pack of 10
Estimated Shipment: 17-05-2024
to see stock.
Add to basket
Description
Description
KECK™ clamps are ground joint clamps used to secure standard ground joint connections in glass apparatus and are characterized by their easy handling and excellent resilience. The clamps are available in different materials (metal or polyoxymethylene).
  • For conical joints
  • Dimensionally stable with particularly good spring properties
  • Easy, safe and quick to set up
  • Increased safety in the laboratory as the glass surface is not scratched
  • Marking of the individual sizes for better differentiation
Specifications
Specifications
10
Secure Glass-to-Glass Joints
NS 12
POM
Secure glass-to-glass joints
Violet