missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences™ KECK™ Clip for Conical Joints, POM, KC

Product Code. 10039920 Shop All DWK Life Sciences Products
Change view
Click to view available options
Size:
NS 10
NS 12
NS 14
NS 19
NS 24
NS 29
NS 34
NS 40
NS 45
Color:
Amber
Blue
Green
Orange
Purple
Red
Yellow
Unit Size:
Each
Pack of 100
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Color unitSize
10039920 NS 10 Green Each
15884678 NS 14 Yellow Pack of 100
15894678 NS 19 Blue Pack of 100
15804688 NS 24 Green Pack of 100
15814688 NS 29 Red Pack of 100
15874678 NS 12 Purple Pack of 100
10468762 NS 34 Orange Each
15981571 NS 40 Yellow Pack of 100
10633982 NS 45 Amber Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Product Code. 10039920 Supplier DWK Life Sciences™ Supplier No. 286336109

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

KECK™ conical ground clamps for a safe ground joint.

KECK™ POM (polyoxymethylene) conical ground joint clamps are easy, safe and quick to attach and are characterized by their dimensional stability and good spring properties. In contrast to conventional wire clamps, the glass surface is not damaged when it is attached. This increases safety in the laboratory. The sizes can be easily distinguished from each other by the different colours.

  • Excellent dimensional stability and good spring properties
  • No damage to the glass surface during attachment unlike conventional wire clamps
  • Easy, safe and quick to put on and take off
  • Colour coding per size
TRUSTED_SUSTAINABILITY

Specifications

Quantity 100
Material POM
For Use With (Equipment) Securing of Conical Ground Joints
For Use With (Application) Securing of conical ground joints
Size NS 10
Color Green
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.