Learn More
DWK Life Sciences™ KECK™ Adapter KA, complete, from hose connection glass to connection plastic
Shop All DWK Life Sciences ProductsDescription
KECK™ adapters prevent glass breakage on olives and protect fingers and hands from injuries. The clamping ring, O-ring and screw cap are permanently screwed to the glass olive. The hose part can be connected quickly, securely and tightly to the threaded cap with just 1/4 turn and can also be loosened again just as quickly.
- Quick connection and disconnection of the hose part to the threaded cap with only 1/4 turn
- permanent screw connection with the glass nozzle
- KECK™ adapters prevent glass breakage on nozzles and protect fingers and hands from injuries
Specifications
Specifications
| Quantity | 10 |
| For Use With (Equipment) | Connection of a Hose Piece with a Threaded Cap |
| For Use With (Application) | Connection of a hose piece with a threaded cap |
| Size | RD 14, diameter 9.0 mm |