missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KDM3B Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | KDM3B |
|---|---|
| Concentration | 0.41 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16859384
|
Proteintech
19915-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16849384
|
Proteintech
19915-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Histone demethylase that specifically demethylates ′Lys-9′ of histone H3, thereby playing a central role in histone code. Demethylation of Lys residue generates formaldehyde and succinate. May have tumor suppressor activity.Specifications
| KDM3B | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Mouse, Human | |
| Q6ZPY7, Q7LBC6 | |
| 277250, 51780 | |
| KDM3B Fusion Protein Ag13795 | |
| Primary | |
| -20°C | |
| KDM3B |
| 0.41 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| 5qNCA, C5orf7, JHDM2B, JMJD1B, KDM3B, KIAA1082, lysine (K) demethylase 3B, Lysine specific demethylase 3B, NET22, Nuclear protein 5qNCA | |
| KDM3B | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title