missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KDM3B Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19915-1-AP-150UL
This item is not returnable.
View return policy
Description
Histone demethylase that specifically demethylates ′Lys-9′ of histone H3, thereby playing a central role in histone code. Demethylation of Lys residue generates formaldehyde and succinate. May have tumor suppressor activity.Specifications
| KDM3B | |
| Polyclonal | |
| Unconjugated | |
| KDM3B | |
| 5qNCA, C5orf7, JHDM2B, JMJD1B, KDM3B, KIAA1082, lysine (K) demethylase 3B, Lysine specific demethylase 3B, NET22, Nuclear protein 5qNCA | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 277250, 51780 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.41 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q6ZPY7, Q7LBC6 | |
| KDM3B | |
| KDM3B Fusion Protein Ag13795 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction