Learn More
KD Scientific Stainless-Steel Syringes
Durable, electron-beam construction withstands high force, high pressure applications
Brand:  KD Scientific 780808
Additional Details : Weight : 0.17000kg
 View more versions of this product
Product Code. 15164174
  • £905.00 / Each
Estimated Shipment: 25-07-2024
to see stock.
Add to basket
Description
Description
Eliminates hazards of glass syringe breakage.
  • Compatible with most syringe pumps
  • Fully autoclavable for reuse
  • Adaptable to Luer Lock or Swagelok™ fittings
  • Resistant to most chemicals
Specifications
Specifications
20 mL
22/25 in.
19 mm
Stainless Steel (316)
1500 psi
3/4 in.
22.3 mm
Syringe Pump
Swagelok 1/8 in. Syringe