missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KD Scientific Stainless-Steel Syringes
Durable, electron-beam construction withstands high force, high pressure applications
Brand: KD Scientific 780802
Code : A0
Additional Details : Weight : 0.35720kg
Description
Eliminates hazards of glass syringe breakage.
- Compatible with most syringe pumps
- Fully autoclavable for reuse
- Adaptable to Luer Lock or Swagelok™ fittings
- Resistant to most chemicals
Specifications
8 mL | |
1/2 in. | |
9.3 mm | |
Stainless Steel (316) | |
1500 psi |
37/100 in. | |
12.7 mm | |
Syringe Pump | |
Swagelok 1/16 in. Syringe |