missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KCTD15 Mouse anti-Human, Mouse, Clone: 2B11, Novus Biologicals™
Mouse Monoclonal Antibody
£232.00 - £358.00
Specifications
| Antigen | KCTD15 |
|---|---|
| Clone | 2B11 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:100 |
| Applications | Western Blot, ELISA, Immunocytochemistry, Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18284943
|
Novus Biologicals
NBP2-60232-50UL |
50 μL |
£232.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18259991
|
Novus Biologicals
NBP2-60232-100UL |
100 μL |
£358.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
KCTD15 Monoclonal specifically detects KCTD15 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| KCTD15 | |
| 1 mg/mL | |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Human, Mouse | |
| BTB/POZ domain-containing protein KCTD15, MGC25497, MGC2628, potassium channel tetramerisation domain containing 15 | |
| KCTD15 | |
| IgG2b κ | |
| Protein G purified |
| 2B11 | |
| Western Blot 1:1000, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:100 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 79047 | |
| Recombinant human KCTD15(1-159aa) purified from E. coli. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
RUO
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title