missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KCTD15 Mouse anti-Human, Mouse, Clone: 2B11, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-60232-100UL
This item is not returnable.
View return policy
Description
KCTD15 Monoclonal specifically detects KCTD15 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| KCTD15 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:100 | |
| BTB/POZ domain-containing protein KCTD15, MGC25497, MGC2628, potassium channel tetramerisation domain containing 15 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 79047 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2b κ |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| 2B11 | |
| Unconjugated | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| KCTD15 | |
| Recombinant human KCTD15(1-159aa) purified from E. coli. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
RUO
Spot an opportunity for improvement?Share a Content Correction