missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KCNMB4 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£359.00
Specifications
| Antigen | KCNMB4 |
|---|---|
| Dilution | Western Blot 1.0 ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
KCNMB4 Polyclonal specifically detects KCNMB4 in Human samples. It is validated for Western Blot.Specifications
| KCNMB4 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| BK channel subunit beta-4, BKbeta4, calcium-activated potassium channel subunit beta-4, Calcium-activated potassium channel, subfamily M subunit beta-4, Charybdotoxin receptor subunit beta-4, hbeta4, K(VCA)beta-4, large conductance calcium-dependent potassium ion channel beta 4 subunit, Maxi K channel subunit beta-4, potassium large conductance calcium-activated channel, subfamily M, beta member4, slo-beta-4 | |
| The immunogen is a synthetic peptide directed towards the middle region of human KCNMB4 (NP_055320). Peptide sequence TCGADCRGTSQYPCVQVYVNNSESNSRALLHSDEHQLLTNPKCSYIPPCK | |
| Affinity purified |
| Western Blot 1.0 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS buffer, 2% sucrose | |
| 27345 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title