missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KCNJ1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£133.00 - £396.00
Specifications
| Antigen | KCNJ1 |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 10-50 ug/ml, Immunohistochemistry 10-50 ug/ml, Immunohistochemistry-Paraffin 10-50 ug/ml, Immunohistochemistry-Frozen 10-50 ug/ml |
| Applications | Western Blot, Immunohistochemistry (Frozen), Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18255005
|
Novus Biologicals
NBP1-30792-0.1MG |
0.1 mg |
£133.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18246054
|
Novus Biologicals
NBP1-30792 |
0.5 mg |
£396.00
0.50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
KCNJ1 Polyclonal specifically detects KCNJ1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| KCNJ1 | |
| Western Blot 10-50 ug/ml, Immunohistochemistry 10-50 ug/ml, Immunohistochemistry-Paraffin 10-50 ug/ml, Immunohistochemistry-Frozen 10-50 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| Sterilized water with No Preservative | |
| ATP-regulated potassium channel ROM-K, ATP-sensitive inward rectifier potassium channel 1, inwardly rectifying K+ channel, Kir1.1, potassium inwardly-rectifying channel, subfamily J, member 1, ROMK1inwardly rectifying subfamily J member 1 | |
| KCNJ1 | |
| Primary | |
| Human. Predicted cross-reactivity based on sequence identity: Mouse (92%), Rat (92%). |
| LYOPH | |
| Western Blot, Immunohistochemistry (Frozen), Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| P48048 | |
| 3758 | |
| IgG | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title