missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KCNJ1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-30792
This item is not returnable.
View return policy
Description
KCNJ1 Polyclonal specifically detects KCNJ1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| KCNJ1 | |
| Polyclonal | |
| Unconjugated | |
| Sterilized water with No Preservative | |
| ATP-regulated potassium channel ROM-K, ATP-sensitive inward rectifier potassium channel 1, inwardly rectifying K+ channel, Kir1.1, potassium inwardly-rectifying channel, subfamily J, member 1, ROMK1inwardly rectifying subfamily J member 1 | |
| Rabbit | |
| Ammonium sulfate precipitation | |
| RUO | |
| 3758 | |
| Reconstitute 0.5 mg size in 0.5 ml of sterile water. Centrifuge to remove any insoluble material. Glycerol may be added (1:1) for additional stability. Please note the 0.1 mg size is provided in reconstituted format. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| LYOPH | |
| Western Blot 10-50 ug/ml, Immunohistochemistry 10-50 ug/ml, Immunohistochemistry-Paraffin 10-50 ug/ml, Immunohistochemistry-Frozen 10-50 ug/ml | |
| P48048 | |
| KCNJ1 | |
| A synthetic peptide from the cytoplasmic domain of human KCNJ1 conjugated to blue carrier protein was used as the antigen. | |
| 0.5 mg | |
| Primary | |
| Human. Predicted cross-reactivity based on sequence identity: Mouse (92%), Rat (92%). | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction