missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KCNA1 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
£119.00 - £295.00
Specifications
| Antigen | KCNA1 |
|---|---|
| Clone | 1D6B9 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30236319
|
Proteintech
68008-1-IG-20UL |
20 μL |
£135.00 £119.00 / 20µL Save £16.00 11% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30236611
|
Proteintech
68008-1-IG-150UL |
150 μL |
£349.00 £295.00 / 150µL Save £54.00 15% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The KCNA1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human KCNA1. This antibody recognizes human, pig, mouse, rat, rabbit antigen. The KCNA1 antibody has been validated for the following applications: WB, ELISA analysis.Specifications
| KCNA1 | |
| 1000 μg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| 3736 | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. |
| 1D6B9 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Mouse | |
| Human, Pig, Mouse, Rat, Rabbit | |
| AEMK; EA1; HBK1; HUK1; KV1.1; MBK1; MGC126782; MGC138385; MK1; RBK1 | |
| IgG2a | |
| Protein A purification | |
| Monoclonal Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title