missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KCNA1 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68008-1-IG-20UL
This item is not returnable.
View return policy
Description
The KCNA1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human KCNA1. This antibody recognizes human, pig, mouse, rat, rabbit antigen. The KCNA1 antibody has been validated for the following applications: WB, ELISA analysis.Specifications
| KCNA1 | |
| Monoclonal | |
| 1000 μg/mL | |
| AEMK; EA1; HBK1; HUK1; KV1.1; MBK1; MGC126782; MGC138385; MK1; RBK1 | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human, Pig, Mouse, Rat, Rabbit | |
| 75 kDa | |
| Liquid |
| Western Blot, ELISA | |
| 1D6B9 | |
| Unconjugated | |
| Mouse | |
| Protein A purification | |
| RUO | |
| 3736 | |
| Store at -20°C. | |
| Monoclonal Antibody | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction