missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KChIP2 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£359.00
Specifications
| Antigen | KChIP2 |
|---|---|
| Dilution | Western Blot 1.0 ug/ml, Immunohistochemistry |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
KChIP2 Polyclonal specifically detects KChIP2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
| KChIP2 | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Vision | |
| PBS buffer, 2% sucrose | |
| 30819 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot 1.0 ug/ml, Immunohistochemistry | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| A-type potassium channel modulatory protein 2, cardiac voltage gated potassium channel modulatory subunit, Cardiac voltage-gated potassium channel modulatory subunit, DKFZp566L1246, KChIP2, KCHIP2Kv channel-interacting protein 2, Kv channel interacting protein 2, MGC17241, potassium channel interacting protein 2, Potassium channel-interacting protein 2 | |
| The immunogen is a synthetic peptide directed towards the N terminal region of human KChIP2 (NP_775285). Peptide sequence QLTDSVDDEFELSTVCHRPEGLEQLQEQTKFTRKELQVLYRGFKNPGALS | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title