missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ KB SRC 4
GREENER_CHOICE

Product Code. 30001606
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 30001606

Brand: Tocris Bioscience™ 4660/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective c-Src inhibitor

Potent and selective c-Src inhibitor (Ki = 44 nM); does not inhibit c-Abl at concentrations up to 125 μM. Selective between Src family members (Kd values are 86, 160, 240, 720, 3200, 4400, and >40,000 nM for c-Src, Lck, Fgr, Yes, Lyn, Hck and Fyn respectively). Significantly inhibits cell growth in 4T1 mammary carcinoma tumor cells.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1380088-03-8
Molecular Formula C32H23ClN8

Specifications

Quantity 10 mg
Formula Weight 555.03
Purity >98%
Molecular Weight (g/mol) 555.042
Chemical Name or Material KB SRC 4
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.