accessibility menu, dialog, popup

UserName

CAS RN 1380088-03-8

NNNNNClNNNH2

CAS RN 1380088-03-8

IUPAC Name: 1-{3-[1-({[1,1'-biphenyl]-3-yl}methyl)-1H-1,2,3-triazol-5-yl]phenyl}-3-(4-chlorophenyl)-1H-pyrazolo[3,4-d]pyrimidin-4-amine
Synonyms: 1-[3-(3-{[1,1'-biphenyl]-3-ylmethyl}-1,2,3-triazol-4-yl)phenyl]-3-(4-chlorophenyl)pyrazolo[3,4-d]pyrimidin-4-amine
Molecular Weight (g/mol): 555.04
Molecular Formula: C32H23ClN8
InChi Key: UHIZYQVRKSWIFO-UHFFFAOYSA-N
SMILES: NC1=C2C(=NC=N1)N(N=C2C1=CC=C(Cl)C=C1)C1=CC=CC(=C1)C1=CN=NN1CC1=CC=CC(=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results