missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KAT4/TBP Associated Factor 1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£359.00
Specifications
| Antigen | KAT4/TBP Associated Factor 1 |
|---|---|
| Dilution | Western Blot 1.0 ug/ml, Chromatin Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
Description
KAT4/TBP Associated Factor 1 Polyclonal specifically detects KAT4/TBP Associated Factor 1 in Human samples. It is validated for Western Blot, Chromatin Immunoprecipitation.Specifications
| KAT4/TBP Associated Factor 1 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| 250kDa, BA2Rdystonia 3 (with Parkinsonism), CCG1TATA box binding protein (TBP)-associated factor, RNA polymerase II, A, 250kD, CCGS, Cell cycle gene 1 protein, cell cycle, G1 phase defect, complementation of cell cycle block, G1-to-S, DYT3/TAF1, EC 2.7.11, EC 2.7.11.1, KAT4, NSCL2, OF, P250, TAF(II)250, TAF1 RNA polymerase II, TATA box binding protein (TBP)-associated factor, TAF2AN-TAF1, TAFII-250, TAFII250DYT3, TBP-associated factor 250 kDa, transcription factor TFIID p250 polypeptide, Transcription initiation factor TFIID 250 kDa subunit, transcription initiation factor TFIID subunit 1, XDP | |
| The immunogen is a synthetic peptide directed towards the middle region of human KAT4/TBP Associated Factor 1 (NP_004597). Peptide sequence MMSYEGDGGEASHGLEDSNISYGSYEEPDPKSNTQDTSFSSIGGYEVSEE | |
| Affinity purified |
| Western Blot 1.0 ug/ml, Chromatin Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Protein Kinase | |
| PBS buffer, 2% sucrose | |
| 6872 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title