missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KAT3B/p300 Antibody (RW105) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 9 publications
£189.00 - £423.00
Specifications
| Antigen | KAT3B/p300 |
|---|---|
| Clone | RW105 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:250-1:500, Flow Cytometry, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:10-1:500, In vitro assay reported in scientific literature (PMID 25728767), Electron Microscopy reported in scientific literature (PMID 25728767), Flow (Intracellular) 1ug/mL, Chromatin Immunoprecipitation (ChIP), KnockDown Validated Validated for KnockDown from a verified customer review. |
| Applications | Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunoprecipitation, In vitro Assay |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18066834
|
Novus Biologicals
NB100-616SS |
0.025 mg |
£189.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18706723
|
Novus Biologicals
NB100-616 |
0.1 mg |
£423.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
KAT3B/p300 Monoclonal specifically detects KAT3B/p300 in Human, Mouse, Rat, Mustelid, Primate samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, In vitro assay, Electron Microscopy, Flow (Intracellular).Specifications
| KAT3B/p300 | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunoprecipitation, In vitro Assay | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Autophagy, Breast Cancer, Cancer, Cell Cycle and Replication, Cellular Markers, Chromatin Research, DNA Repair, HIF Target Genes, Hypoxia, Stem Cell Markers, Transcription Factors and Regulators, Tumor Suppressors, Wnt Signaling Pathway | |
| E1A binding protein p300, E1A-associated protein p300, E1A-binding protein, 300kD, EC 2.3.1, EC 2.3.1.48, histone acetyltransferase p300, KAT3B, P300, p300 HAT, RSTS2 | |
| EP300 | |
| IgG1 κ | |
| Protein G purified | |
| This is specific for p300 protein. This recognizes residues 1921-2023. |
| RW105 | |
| Western Blot 1:250-1:500, Flow Cytometry, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:10-1:500, In vitro assay reported in scientific literature (PMID 25728767), Electron Microscopy reported in scientific literature (PMID 25728767), Flow (Intracellular) 1ug/mL, Chromatin Immunoprecipitation (ChIP), KnockDown Validated Validated for KnockDown from a verified customer review. | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat, Mustelid, Primate | |
| 2033 | |
| Fusion protein containing residues 1572-2371 of human KAT3B/p300. [UniProt# Q0947] | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title