missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KAT3B/p300 Antibody (RW105) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 9 publications
Brand: Novus Biologicals NB100-616SS
This item is not returnable.
View return policy
Description
KAT3B/p300 Monoclonal specifically detects KAT3B/p300 in Human, Mouse, Rat, Mustelid, Primate samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, In vitro assay, Electron Microscopy, Flow (Intracellular).
Specifications
| KAT3B/p300 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:250-1:500, Flow Cytometry, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:10-1:500, In vitro assay reported in scientific literature (PMID 25728767), Electron Microscopy reported in scientific literature (PMID 25728767), Flow (Intracellular) 1ug/mL, Chromatin Immunoprecipitation (ChIP), KnockDown Validated Validated for KnockDown from a verified customer review. | |
| EP300 | |
| Fusion protein containing residues 1572-2371 of human KAT3B/p300. [UniProt# Q0947] | |
| 0.025 mg | |
| Apoptosis, Autophagy, Breast Cancer, Cancer, Cell Cycle and Replication, Cellular Markers, Chromatin Research, DNA Repair, HIF Target Genes, Hypoxia, Stem Cell Markers, Transcription Factors and Regulators, Tumor Suppressors, Wnt Signaling Pathway | |
| 2033 | |
| Human, Mouse, Rat, Mustelid, Primate | |
| Purified |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunoprecipitation, In vitro Assay | |
| RW105 | |
| Unconjugated | |
| E1A binding protein p300, E1A-associated protein p300, E1A-binding protein, 300kD, EC 2.3.1, EC 2.3.1.48, histone acetyltransferase p300, KAT3B, P300, p300 HAT, RSTS2 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| This is specific for p300 protein. This recognizes residues 1921-2023. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction