missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Karmali Selective Agar, Blood-Free

Product Code. 13295349
Change view
Click to view available options
Format:
90mm Monoplate
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Format Unit Size
13295349 90mm Monoplate Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13295349 Supplier Thermo Scientific™ Supplier No. PO5041A
  • ONLINE EXCLUSIVE PRICE
    £10.85
    £9.63 / Pack of 10
    Save £1.22
    11% Off
 Restricted Item
Estimated Shipment: 31-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Isolate and identify Campylobacter jejuni and Campylobacter coli from clinical specimens with this blood-free medium.

Isolate and identify Campylobacter jejuni and Campylobacter coli from clinical specimens using blood-free Thermo Scientific™ Karmali Selective Agar.

Based on the formulation described by Karmali, this formulation is supplemented with vancomycin to inhibit the growth of Gram-positive organisms. Campylobacter growth is recognized as shiny grey colonies.

Campylobacter
infections are among the most common bacterial infections, producing both gastrointestinal and systemic illnesses. Campylobacter jejuni is the most commonly reported bacterial cause of foodborne infection in the United States. Campylobacter are small, non-sporeforming, helical, curved gram-negative rods that require a microaerophilic environment and cannot tolerate drying.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance According ISO 10272
Color Jet black, opaque
Content And Storage 2° to 12°C, in the dark
Description Karmali Selective Agar, Blood-Free
Format 90mm Monoplate
Manufacturing Origin Germany
Product Type Agar Plate
Fill Weight 17g ± 5%
Packaging Type Flow wrapped - single layer
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.