missing translation for 'onlineSavingsMsg'
Learn More

Poeppelmann™ KAPSTO™ GPN 610 Universal Protection Caps/Plugs

Product Code. p-8003241
Click to view available options
Diameter (Metric) Outside:
8.3 dia. x 10.0mmH
Diameter (Metric) Inside:
9.9 dia. x 10.0mmH
Height (Metric):
15 mm
Unit Size:
Pack of 100

Product Code. 10155584

Brand: Poeppelmann™ 61000060000

Only null left
Add to basket
Add to basket

Excellent for closing off bore holes and internal threads, or to protect studs and external threads. Poeppelmann™ KAPSTO™ GPN 610 Universal Protection Caps/Plugs are manufactured from yellow PE-LD plastic.

  • Universal use as a plug or cap
  • Acc. with VDA Band 19 and ISO 16232
  • Tapered design ideal for accomodating tolerances

Specifications

Color Yellow
Diameter (Metric) Inside 9.9 dia. x 10.0mmH
For Use With (Application) Used universally as plug or cap, Tapered design is ideal for accommHodating tolerances
Height (Metric) 15 mm
Length (Metric) 10.1 mm
Quantity 100 Pack
Diameter (Metric) Outside 8.3 dia. x 10.0mmH
Material Polyethylene
Type Closing Plug
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.