missing translation for 'onlineSavingsMsg'
Learn More

Poeppelmann™ KAPSTO™ GPN 610 Universal Protection Caps/Plugs

Product Code. 10155584
Change view
Click to view available options
Diameter (Metric) Outside:
8.3 dia. x 10.0mmH
Diameter (Metric) Inside:
9.9 dia. x 10.0mmH
Height (Metric):
15 mm
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Outside Diameter (Metric) Inside Height (Metric) Unit Size
10155584 8.3 dia. x 10.0mmH 9.9 dia. x 10.0mmH 15 mm Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10155584 Supplier Poeppelmann™ Supplier No. 61000060000
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

Excellent for closing off bore holes and internal threads, or to protect studs and external threads. Poeppelmann™ KAPSTO™ GPN 610 Universal Protection Caps/Plugs are manufactured from yellow PE-LD plastic.

  • Universal use as a plug or cap
  • Acc. with VDA Band 19 and ISO 16232
  • Tapered design ideal for accomodating tolerances

Specifications

Color Yellow
Diameter (Metric) Inside 9.9 dia. x 10.0mmH
For Use With (Application) Used universally as plug or cap, Tapered design is ideal for accommHodating tolerances
Height (Metric) 15 mm
Length (Metric) 10.1 mm
Quantity 100 Pack
Diameter (Metric) Outside 8.3 dia. x 10.0mmH
Material Polyethylene
Type Closing Plug
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.