missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Kanamycin Aesculin Azide Selective Medium

Product Code. 10422133
Change view
Click to view available options
Format:
90mm Monoplate
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Format Unit Size
10422133 90mm Monoplate Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10422133 Supplier Thermo Scientific™ Supplier No. PO5059A
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Isolate, differentiate and enumerate enterococci from food samples using a selective microbiological medium which contains a number of different selective agents.

Isolate, differentiate and enumerate enterococci from food samples using Thermo Scientific™ Kanamycin Aesculin Azide Selective Medium, a selective microbiological media which contains a number of different selective agents.

Addition of the selective inhibitory components kanamycin sulphate and sodium azide, in addition to an indicator system, detect the growth of aesculin-hydrolysing streptococci. These organisms produce black zones around the colonies due to the formation of black iron phenolic compounds derived from aesculin-hydrolysis products and ferrous iron.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY

Specifications

Color Green beige, transparent
Content And Storage 6°C to 12°C
Description Kanamycin Aesculin Azide Selective Medium
Format 90mm Monoplate
Manufacturing Origin Germany
Product Type Agar Plate
Fill Weight 17g ± 5%
Packaging Type Flow Wrapped - Single Layer
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.