missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Kanamycin Aesculin Azide Agar

Product Code. 12902598
This item is not returnable. View return policy

Product Code. 12902598

Brand: Thermo Scientific™ PO0173A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Detect and isolate enterococci in food stuffs with this selective medium.

Detect and isolate Enterococci in foodstuffs with Thermo Scientific™ Oxoid™ Kanamycin Aesculin Azide Agar. Due to the presence of selective inhibitory components kanamycin sulphate and sodium azide medium provides selectivity against non-target organisms. Aesculin and ferric ammonium citrate together form the indicator system to detect Enterococci, which form black zones around the colonies. The black zones are produced from the formation of black iron phenolic compounds derived from aesculin hydrolysis products and ferrous ions.

Enterococci are part of the normal intestinal flora of human and animals. They have been long recognized as important human pathogens and are becoming increasingly so. Kanamycin Esculin Azide Agar is formulated as per Mossel et al 1, 2 to detect Enterococci in foodstuffs. Mossel et al3 used it for the dip slide technique for bacteriological monitoring of foods.

  • Selective: due to the presence of kanamycin sulphate and sodium azide medium provides selectivity against non-target organisms.
  • Easy to read: aesculin-hydrolyzing Streptococci form black zones around the colonies.
  • Can be used successfully for the isolation of glycopeptide resistant enterococci from clinical specimens and foods4,5
  • Ready to use: convenience of prepared media.

Members of the genus Enterococcus were classified as group D Streptococcus until 1984, when genomic DNA analysis indicated a separate genus classification would be appropriate.6

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Color Straw / Green with blue hue, Clear
Content And Storage 2°C to 10°C
Description Kanamycin Aesculin Azide Agar
Format 90mm Monoplate
Manufacturing Origin United Kingdom
Product Type Agar
Fill Weight 19.5g ± 1.0g
Packaging Type Ten 90mm plates wrapped in a single cellulose-based film wrap
Quantity 10/Pk.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.