missing translation for 'onlineSavingsMsg'
Learn More

Kallikrein 6/Neurosin Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£178.00 - £255.00

Specifications

Antigen Kallikrein 6/Neurosin
Dilution Western Blot 1:500-1:1000, ELISA 1:25000-1:50000
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18624052
View Documents
Novus Biologicals
NBP2-98247-100ul
100 μg
£255.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18640265
View Documents
Novus Biologicals
NBP2-98247-50ul
50 μg
£178.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Kallikrein 6/Neurosin Polyclonal antibody specifically detects Kallikrein 6/Neurosin in Mouse samples. It is validated for Western Blot, ELISA
Specifications

Specifications

Kallikrein 6/Neurosin
Western Blot, ELISA
Unconjugated
Rabbit
Cancer, Cell Biology, Cellular Markers, Endothelial Cell Markers, Extracellular Matrix, Neurodegeneration, Neuroscience, Signal Transduction
0.2 um filtered solution in PBS
5653
IgG
Antigen and protein A Affinity-purified
Western Blot 1:500-1:1000, ELISA 1:25000-1:50000
Polyclonal
Purified
RUO
Mouse
Bssp, EC 3.4.21, EC 3.4.21.-, hK6, kallikrein 6 (neurosin, zyme), kallikrein-6, kallikrein-related peptidase 6, Klk7, neurosin, Protease M, protease, serine, 18, PRSS18, PRSS9MGC9355, Serine protease 18, Serine protease 9, SP59, Zyme
Produced in rabbits immunized with purified, recombinant Mouse Kallikrein 6/Neurosin (Accession#: NP_035307.2; Met 8-Leu 253)
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.