missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kallikrein 6/Neurosin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98247-100ul
This item is not returnable.
View return policy
Description
Kallikrein 6/Neurosin Polyclonal antibody specifically detects Kallikrein 6/Neurosin in Mouse samples. It is validated for Western Blot, ELISA
Specifications
| Kallikrein 6/Neurosin | |
| Polyclonal | |
| Western Blot 1:500-1:1000, ELISA 1:25000-1:50000 | |
| Bssp, EC 3.4.21, EC 3.4.21.-, hK6, kallikrein 6 (neurosin, zyme), kallikrein-6, kallikrein-related peptidase 6, Klk7, neurosin, Protease M, protease, serine, 18, PRSS18, PRSS9MGC9355, Serine protease 18, Serine protease 9, SP59, Zyme | |
| Produced in rabbits immunized with purified, recombinant Mouse Kallikrein 6/Neurosin (Accession#: NP_035307.2; Met 8-Leu 253) | |
| 100 μg | |
| Cancer, Cell Biology, Cellular Markers, Endothelial Cell Markers, Extracellular Matrix, Neurodegeneration, Neuroscience, Signal Transduction | |
| 5653 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ELISA | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction