missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kallikrein 4/Prostase/EMSP1 Antibody (KLK4/8943), Novus Biologicals™
Mouse Monoclonal Antibody
£195.00 - £368.00
Specifications
| Antigen | Kallikrein 4/Prostase/EMSP1 |
|---|---|
| Clone | KLK4/8943 |
| Concentration | 0.2 mg/ml |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30300386
|
Novus Biologicals
NBP3-44627-20ug |
20 μg |
£195.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30293957
|
Novus Biologicals
NBP3-44627-100ug |
100 μg |
£368.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Kallikrein 4/Prostase/EMSP1 Monoclonal antibody specifically detects Kallikrein 4/Prostase/EMSP1 in Human samples. It is validated for ImmunohistochemistrySpecifications
| Kallikrein 4/Prostase/EMSP1 | |
| 0.2 mg/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| AI2A1, ARM1, EC 3.4.21, EC 3.4.21.-, EMSP, EMSP1MGC116827, Enamel matrix serine proteinase 1, kallikrein 4 (prostase, enamel matrix, prostate), kallikrein-4, Kallikrein-like protein 1, kallikrein-related peptidase 4, KLK-L1MGC116828, Prostase, PRSS17enamel matrix serine protease 1, PSTSandrogen-regulated message 1, Serine protease 17 | |
| Recombinant fragment of human Kallikrein 4/Prostase/EMSP1 protein (around aa 1-200) (Uniprot: Q9Y5K2). Exact sequence is proprietary. | |
| Primary | |
| Store at 4⪚C. |
| KLK4/8943 | |
| Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Cancer, Tumor Suppressors | |
| 10 mM PBS with 0.05% BSA | |
| 9622 | |
| IgG1 κ | |
| Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title