Learn More
Kallikrein 4/Prostase/EMSP1 Antibody (KLK4/8943), Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-44627-20ug
 View more versions of this product
Product Code. 30300386
  • £195.00 / 20µg
Description
Kallikrein 4/Prostase/EMSP1 Monoclonal antibody specifically detects Kallikrein 4/Prostase/EMSP1 in Human samples. It is validated for Immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Kallikrein 4/Prostase/EMSP1
Monoclonal
0.2 mg/ml
10 mM PBS with 0.05% BSA
Mouse
Protein G purified
RUO
Primary
Human
Purified
Immunohistochemistry
KLK4/8943
Unconjugated
AI2A1, ARM1, EC 3.4.21, EC 3.4.21.-, EMSP, EMSP1MGC116827, Enamel matrix serine proteinase 1, kallikrein 4 (prostase, enamel matrix, prostate), kallikrein-4, Kallikrein-like protein 1, kallikrein-related peptidase 4, KLK-L1MGC116828, Prostase, PRSS17enamel matrix serine protease 1, PSTSandrogen-regulated message 1, Serine protease 17
Recombinant fragment of human Kallikrein 4/Prostase/EMSP1 protein (around aa 1-200) (Uniprot: Q9Y5K2). Exact sequence is proprietary.
20 μg
Apoptosis, Cancer, Tumor Suppressors
9622
Store at 4⪚C.
IgG1 κ