missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Kaiso Recombinant Rabbit Monoclonal Antibody (24GB3670)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557323
This item is not returnable.
View return policy
Description
Kaiso Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
This gene encodes a transcriptional regulator with bimodal DNA-binding specificity, which binds to methylated CGCG and also to the non-methylated consensusKAISO-binding site TCCTGCNA. The protein contains an N-terminal POZ/BTB domain and 3 C-terminal zinc finger motifs. It recruits the N-CoR repressor complex to promote histone deacetylation and the formation of repressive chromatin structures in target gene promoters. It may contribute to the repression of target genes of the Wnt signaling pathway, and may also activate transcription of a subset of target genes by the recruitment of catenin delta-2 (CTNND2). Its interaction with catenin delta-1 (CTNND1) inhibits binding to both methylated and non-methylated DNA. It also interacts directly with the nuclear import receptor Importin-alpha 2 (also known as karyopherin alpha2 or RAG cohort 1), which may mediate nuclear import of this protein. Alternatively spliced transcript variants encoding the same protein have been identified.
Specifications
| Kaiso | |
| Recombinant Monoclonal | |
| Unconjugated | |
| ZBTB33 | |
| AW260253; E130014G12Rik; Kaiso; kaiso transcription factor; RGD1566309; RP23-322E15.9; Transcriptional regulator Kaiso; WUGSC:H_DJ525N14.1; Zbtb33; zinc finger and BTB domain containing 33; zinc finger and BTB domain-containing protein 33; zinc finger transcription factor Kaiso; ZNF348; ZNF-kaiso | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 10009, 501506 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB3670 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q86T24 | |
| ZBTB33 | |
| A synthesized peptide derived from human KAISO (aa 620-672). | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction