missing translation for 'onlineSavingsMsg'
Learn More

Kaempferol, TRC

Product Code. 30040431
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30040431 50 mg 50mg
18072230 100 mg 100mg
18031440 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30040431 Supplier Toronto Research Chemicals Supplier No. TRCK10000050MG
 Restricted Item
Estimated Shipment: 22-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 520-18-3
Molecular Formula C15 H10 O6
Molecular Weight (g/mol) 286.24
Synonym Flavone, 3,4',5,7-tetrahydroxy- (7CI,8CI), 3,5,7-Trihydroxy-2-(4-hydroxyphenyl)-4H-1-benzopyran-4-one, 3,4',5,7-Tetrahydroxyflavone, 3,5,7,4'-Tetrahydroxyflavone, 3'-Deoxyquercetin, 5,7,4'-Trihydroxyflavonol, C.I. 75640, CSU 2056, Indigo Yellow, Kaemferol, Kaempferol, Kaempherol, Kampcetin, Kempferol, NSC 407289, NSC 656277, Nimbecetin, Pelargidenolon, Pelargidenon, Populnetin, Rhamnolutein, Rhamnolutin, Robigenin, Swartziol, Trifolitin
IUPAC Name 3,5,7-trihydroxy-2-(4-hydroxyphenyl)chromen-4-one
SMILES OC1=C(Oc2cc(O)cc(O)c2C1=O)c3ccc(O)cc3

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C15H10O6/c16-8-3-1-7(2-4-8)15-14(20)13(19)12-10(18)5-9(17)6-11(12)21-15/h1-6,16-18,20H
Formula Weight 286.0477
Percent Purity >95%
Chemical Name or Material Kaempferol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.